missing translation for 'onlineSavingsMsg'
Learn More

Current Technologies Dispensers for Bio-Screen™ or Fisherbrand™ BloodBloc™ Wipes

Catalog No. 1441251
Change view
Click to view available options
Dimensions (L x W x H):
3.5 x 9.25 x 3.5 in.
Color:
Clear
Material:
Acrylic
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H) Color Material
1441251 3.5 x 9.25 x 3.5 in. Clear Acrylic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1441251 Supplier Current Technologies Supplier No. ACBH33

Reduce clutter on workbench, counters, trays, carts with either clear acrylic or white cardboard dispensers; sized specifically to store BloodBloc or Bioscreen wipes

For 3 × 3 in. BloodBloc (06-670-1, -36, -49), and Bio-Screen (06-670, 14-412-52C, -52H) biohazard wipes

  • Measures 3W x 3L x 3 in.H
  • For 4 × 4 in. BloodBloc (06-670-35, -50, -5) and Bio-Screen (14-412-52E, -52D, -52G) biohazard wipes
    • Measures 4W × 4L × 5.5 in.H
    • May also be used with full-size HYPE-WIPE™ bleach towelettes, 14-412-56

    Specifications

    Product Type Wiper Holder
    Color Clear
    Dimensions (L x W x H) 3.5 x 9.25 x 3.5 in.
    Length (English) 3.5 in.
    Length (Metric) 8.89 cm
    Width (English) 9.25 in.
    Width (Metric) 2.54 cm
    Height (English) 3.5 in.
    Height (Metric) 8.89 cm
    Material Acrylic
    For Use With (Application) Reduce clutter on workbench, counters, trays, carts
    For Use With (Equipment) For 3 x 3 in. biohazard wipes
    Show More Show Less