missing translation for 'onlineSavingsMsg'
Learn More

Current Technologies Clear Acrylic Holders for HYPE-WIPE™ Towelettes

Catalog No. 22282230
Change view
Click to view available options
Dimensions (L x W x H):
4 x 7.75 x 3.5 in.
4 x 4 x 5.5 in.
For Use With (Equipment):
HYPE-WIPE, 4 x 4 x 5.5 in. Bio-Hazard Wipes
Mini HYPE-WIPES
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H) For Use With (Equipment)
22282230 4 x 4 x 5.5 in. HYPE-WIPE, 4 x 4 x 5.5 in. Bio-Hazard Wipes
1441250 4 x 7.75 x 3.5 in. Mini HYPE-WIPES
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22282230 Supplier Current Technologies Supplier No. AC9512

Reduce clutter on workbench, counters, trays, carts with clear acrylic dispensers for use with HYPE-WIPE™ and Mini HYPE-WIPE™ bleach towelettes

Specifications

Product Type Wiper Dispenser
Color Clear
Dimensions (L x W x H) 4 x 4 x 5.5 in.
Length (English) 4 in.
Length (Metric) 10.16 cm
Width (English) 4 in.
Width (Metric) 114.3 mm
Height (English) 5 in.
Height (Metric) 12.7 cm
Material Acrylic
For Use With (Application) Reduce clutter on workbench, counters, trays, carts
For Use With (Equipment) HYPE-WIPE, 4 x 4 x 5.5 in. Bio-Hazard Wipes
Show More Show Less