Learn More
CurranTaylor™ Scotsman™ Filters and Filter Cartridges for Ice Makers
Catalog No. 09540123
Change view
Click to view available options
Capacity:
25,000 gal.
35,000 gal.
9000 gal.
Product Type:
Filter Cartridge for Model 121
Filter Cartridge for Model 122
Filter Cartridge for Model 123
Large System Filter Package
Medium System Filter Package
Small System Filter Package
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity Product Type
09540123 35,000 gal. Large System Filter Package
09540125 25,000 gal. Filter Cartridge for Model 122
09540121 9000 gal. Small System Filter Package
09540122 25,000 gal. Medium System Filter Package
09540124 9000 gal. Filter Cartridge for Model 121
09540126 35,000 gal. Filter Cartridge for Model 123
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 09540123 Supplier CurranTaylor™ Supplier No. IMF160S
Only null left
Scotsman™ Filters and Filter Cartridges for Ice Makers keeps ice makers cleaner while reducing failures due to lime scale, dirt, and excessive chlorine.
  • IMPACT media package in large cartridges
  • Built in scale inhibition
  • SQC™ (Sanitary Quick Change) cartridge
  • BSPT horizontal inlet and outlet port
  • 0.2 micron media
  • Valve-in-head
  • Optional pressure gauge
Benifits:
  • Higher flow rates, no need for manifold systems or for separate pre-filter and housing
  • Reduces ability of calcium and magnesium to precipitate on evaporator plates as hard scale
  • Allows fast and easy change-outs with 1/4 turn while minimising the possibility of leakage and contamination
  • Allows direct or easily adaptable connections to existing plumbing lines
  • Reduces microbial content in water and protects customers from cyst and 99.9% of common pathogens
  • Combined sediment reduction with chlorine, taste and odour reduction for high dirt loading applications
  • No separate shut off valve required, easy to change using un-skilled staff
  • Easy to monitor filter blockage for identification of time to change filter
  • Independent qualification of performance and guarantee of product safety
Specifications
Certifications/Compliance NSF, FDA CFR-21
Capacity (English) 35,000 gal.
Capacity (Metric) 132,489 L
Product Type Large System Filter Package
For Use With (Application) Provides consistent high quality water for commercial ice by reducing the effects of sediment, chlorine, and odor and scale
For Use With (Equipment) Ice makers
Includes Built-in scale inhibitor, sanitary quick change encapsulated cartridge, 0.5 in. FNPT horizontal inlet and outlet ports, head assembly with integral mounting bracket, pressure gauge and single cartridge filter, NSF performance data sheet
Capacity 35,000 gal.