missing translation for 'onlineSavingsMsg'
Learn More

Cuprous Chloride, Technical, Spectrum™ Chemical

Catalog No. p-7165370
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
Catalog No. Quantity Packaging
18610066 500 g Amber Glass Bottle
18610063 2.5 kg Amber Glass Bottle
18610064 12 kg Poly Pail
18610065 45 kg Fiber Drum
4 options

Catalog No. 18610066

Supplier: Spectrum Chemical Mfg Cor C1442500GM

Only null left

Chemical Identifiers

CAS 7758-89-6

Specifications

CAS Max % 1
Quantity 500 g
Grade Technical
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.