missing translation for 'onlineSavingsMsg'
Learn More

Cupric Sulfate Pentahydrate (Fine Crystals/Certified ACS), Fisher Chemical™

Catalog No. C493500
Change view
Click to view available options
Quantity:
500 g
3 kg
10 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
C493500 500 g Poly Bottle
C4933 3 kg Poly Bottle
C49310 10 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C493500 Supplier Thermo Fisher Scientific Supplier No. FLC493500
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7758-99-8
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.68
MDL Number MFCD00149681
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
Synonym copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
PubChem CID 24463
ChEBI CHEBI:31440
IUPAC Name copper(2+) pentahydrate sulfate
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

Boiling Point 150°C
Melting Point 110°C
Color Blue
pH 3.5 to 4.5
Physical Form Solid
Quantity 500 g
Assay Percent Range 98.0 to 102.0%
UN Number 3077
Formula Weight 249.68
Percent Purity 98.0 to 102.0%
Grade Certified ACS
Identification Pass Test
Packaging Poly Bottle
Vapor Pressure 7.3mmHg at 25°C
Density 2.28g/mL
Chemical Name or Material Cupric Sulfate Pentahydrate
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Harmful if swallowed. May cause irritation of respiratory tract. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:2
Flammability:0
Instability:1
DOTInformation DOT Class 9, : Miscellaneous