missing translation for 'onlineSavingsMsg'
Learn More

Cupric Sulfate, Pentahydrate, Fine Crystal, ACS, 98-102%, Spectrum™ Chemical

Catalog No. 18610051
Change view
Click to view available options
Quantity:
5 g
25 g
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610051 45 kg Poly Bottle
18610045 5 g Poly Bottle
18610046 25 g Poly Bottle
18610047 125 g Poly Bottle
18610048 500 g Poly Bottle
18610049 2.5 kg Poly Bottle
18610050 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18610051 Supplier Spectrum Chemical Mfg Cor Supplier No. C142545KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7758-99-8 | CuH10O9S | 249.68 g/mol

Chemical Identifiers

CAS 7758-99-8
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.68
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
IUPAC Name copper(2+) pentahydrate sulfate
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

CAS Max % 1
Physical Form Crystalline Powder, Needles or Crystals
Quantity 45 kg
Percent Purity 98 to 102%
Grade ACS/Reagent
Packaging Poly Bottle
Chemical Name or Material Cupric Sulfate, Pentahydrate, Fine Crystal

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.