missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cupric Sulfate Pentahydrate, ≥99%, (Crystalline), MP Biomedicals
$155.52 - $155.52
Chemical Identifiers
| CAS | 7758-99-8 |
|---|---|
| Molecular Formula | CuH10O9S |
| Molecular Weight (g/mol) | 249.68 |
| MDL Number | MFCD00149681 |
| InChI Key | JZCCFEFSEZPSOG-UHFFFAOYSA-L |
| Synonym | copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas |
| PubChem CID | 24463 |
| ChEBI | CHEBI:31440 |
| IUPAC Name | copper(2+) pentahydrate sulfate |
| SMILES | O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN19511790
|
MP Biomedicals Inc
0219511790 |
500 g |
Each for $155.52
|
|
|||||
Chemical Identifiers
| 7758-99-8 | |
| 249.68 | |
| JZCCFEFSEZPSOG-UHFFFAOYSA-L | |
| 24463 | |
| copper(2+) pentahydrate sulfate |
| CuH10O9S | |
| MFCD00149681 | |
| copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas | |
| CHEBI:31440 | |
| O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O |
Specifications
| 7758-99-8 | |
| 500 g | |
| CuH10O9S | |
| copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas | |
| O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O | |
| 249.68 | |
| CHEBI:31440 | |
| Cupric Sulfate Pentahydrate |
| Crystalline | |
| ≥99% | |
| MFCD00149681 | |
| JZCCFEFSEZPSOG-UHFFFAOYSA-L | |
| copper(2+) pentahydrate sulfate | |
| 24463 | |
| 249.68 |
Safety and Handling
Recommended Storage : Room Temperature