missing translation for 'onlineSavingsMsg'
Learn More

Cupric Sulfate Pentahydrate, ≥99%, (Crystalline), MP Biomedicals

Catalog No. ICN19511790
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19511790 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19511790 Supplier MP Biomedicals Inc Supplier No. 0219511790

Chemical Identifiers

CAS 7758-99-8
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.68
MDL Number MFCD00149681
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
Synonym copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
PubChem CID 24463
ChEBI CHEBI:31440
IUPAC Name copper(2+) pentahydrate sulfate
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

Physical Form Crystalline
Quantity 500 g
Assay Percent Range ≥99%
Formula Weight 249.68
Chemical Name or Material Cupric Sulfate Pentahydrate
Recommended Storage Room Temperature