missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cupric Sulfate Pentahydrate, 98-102%, (Crystalline/ACS Reagent Grade), MP Biomedicals
Specifications
Specifications
| CAS | 7758-99-8 |
| Physical Form | Crystalline |
| Quantity | 100 g |
| Assay Percent Range | 98-102% |
| Molecular Formula | CuH10O9S |
| MDL Number | MFCD00149681 |
| Synonym | copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas |
| InChI Key | JZCCFEFSEZPSOG-UHFFFAOYSA-L |
| SMILES | O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O |
| IUPAC Name | copper(2+) pentahydrate sulfate |
| Show More |
Safety and Handling
| Recommended Storage | Room Temperature |