missing translation for 'onlineSavingsMsg'
Learn More

Cupric Nitrate Hydrate, MP Biomedicals

$265.97 - $265.97

Chemical Identifiers

CAS 10031-43-3
Molecular Formula CuH6N2O9
Molecular Weight (g/mol) 241.599
InChI Key SXTLQDJHRPXDSB-UHFFFAOYSA-N
Synonym copper ii nitrate trihydrate, copper nitrate trihydrate, cupric nitrate trihydrate, copper ii nitrate hydrate, copper ii nitrate 3-hydrate, copper 2+ trihydrate dinitronate, copper 2+ trihydrate dinitrate, copper ii nitrate trihydrate-cu puratrem
PubChem CID 9837674
IUPAC Name copper;dinitrate;trihydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.[Cu+2]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN15350190
View Documents
MP Biomedicals Inc
0215350190
500 g
Each for $265.97
Only null left
 
Description

Description

Specifications

Chemical Identifiers

10031-43-3
241.599
copper ii nitrate trihydrate, copper nitrate trihydrate, cupric nitrate trihydrate, copper ii nitrate hydrate, copper ii nitrate 3-hydrate, copper 2+ trihydrate dinitronate, copper 2+ trihydrate dinitrate, copper ii nitrate trihydrate-cu puratrem
copper;dinitrate;trihydrate
CuH6N2O9
SXTLQDJHRPXDSB-UHFFFAOYSA-N
9837674
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.[Cu+2]

Specifications

10031-43-3
CuH6N2O9
SXTLQDJHRPXDSB-UHFFFAOYSA-N
copper;dinitrate;trihydrate
9837674
Cupric Nitrate Hydrate
500 g
copper ii nitrate trihydrate, copper nitrate trihydrate, cupric nitrate trihydrate, copper ii nitrate hydrate, copper ii nitrate 3-hydrate, copper 2+ trihydrate dinitronate, copper 2+ trihydrate dinitrate, copper ii nitrate trihydrate-cu puratrem
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.[Cu+2]
241.599
187.6 (anhydrous)
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

SDS
missing translation for 'documents'

missing translation for 'documents'