missing translation for 'onlineSavingsMsg'
Learn More

Cupric Nitrate, Hydrate, Crystal, ACS, 98-102%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor C140525GM

 View more versions of this product

Catalog No. 18610006


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

19004-19-4
25 g
ACS
1
98 to 102%
Poly Bottle

Chemical Identifiers

19004-19-4
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.