missing translation for 'onlineSavingsMsg'
Learn More

Cupric Nitrate, Hydrate, Crystal, ACS, 98-102%, Spectrum™ Chemical

Catalog No. 18610005
Change view
Click to view available options
Quantity:
5 g
25 g
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610005 5 g Amber Glass Bottle
18610006 25 g Poly Bottle
18610007 125 g Poly Bottle
18610008 500 g Poly Bottle
18610009 2.5 kg Poly Bottle
18610010 12 kg Poly Pail
18610011 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18610005 Supplier Spectrum Chemical Mfg Cor Supplier No. C14055GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 19004-19-4

Specifications

CAS Max % 1
Quantity 5 g
Percent Purity 98 to 102%
Grade ACS
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.