missing translation for 'onlineSavingsMsg'
Learn More

Cupric Nitrate Hemipentahydrate, Purified (Copper (II) Nitrate Hydrate), Reagents

Catalog No. 01337306
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337306 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01337306 Supplier Labchem Inc Supplier No. C2189000500A5
Only null left

Cupric Nitrate Hemipentahydrate, Purified (Copper (II) Nitrate Hydrate), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 19004-19-4
Molecular Formula Cu2H10N4O17
Molecular Weight (g/mol) 465.18
InChI Key IZDJJEMZQZQQQQ-UHFFFAOYSA-N
Synonym Copper(II) Nitrate Hemipentahydrate
IUPAC Name dicopper(2+) pentahydrate tetranitrate
SMILES O.O.O.O.O.[Cu++].[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Boiling Point 170°C
Melting Point 114.5°C
Color Blue
Physical Form Solid
Quantity 500 g
Grade Pure
Packaging Poly Bottle
Appearance Blue
Specific Gravity 2.32
Chemical Name or Material Cupric Nitrate Hemipentahydrate, Purified (Copper (II) Nitrate Hydrate)