missing translation for 'onlineSavingsMsg'
Learn More

Cupric Nitrate Hemipentahydrate, Purified (Copper (II) Nitrate Hydrate), Reagents

Catalog No. 01337307
Change view
Click to view available options
Quantity:
100 mL
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337307 100 mL
01337306 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337307 Supplier Labchem Inc Supplier No. C2189000100A6

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cupric Nitrate Hemipentahydrate, Purified (Copper (II) Nitrate Hydrate), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 19004-19-4
Molecular Formula Cu2H10N4O17
Molecular Weight (g/mol) 465.18
InChI Key IZDJJEMZQZQQQQ-UHFFFAOYSA-N
Synonym Copper(II) Nitrate Hemipentahydrate
IUPAC Name dicopper(2+) pentahydrate tetranitrate
SMILES O.O.O.O.O.[Cu++].[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Boiling Point 170°C
Melting Point 114.5°C
Color Blue
Physical Form Solid
Quantity 100 mL
Grade Pure
Packaging Poly Bottle
Appearance Blue
Specific Gravity 2.32
Chemical Name or Material Cupric Nitrate Hemipentahydrate, Purified (Copper (II) Nitrate Hydrate)