missing translation for 'onlineSavingsMsg'
Learn More

Cupric Chloride Dihydrate, Purified (Copper (II) Chloride Dihydrate), Reagents

Catalog No. 01337309
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337309 100 g
01337308 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337309 Supplier Labchem Inc Supplier No. C2188500100C2

Cupric Chloride Dihydrate, Purified (Copper (II) Chloride Dihydrate), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 10125-13-0
Molecular Formula Cl2CuH4O2
Molecular Weight (g/mol) 170.48
MDL Number MFCD00149674
InChI Key MPTQRFCYZCXJFQ-UHFFFAOYSA-L
Synonym Copper(II) Chloride Dihydrate
IUPAC Name copper(2+) dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].[Cu++]

Specifications

Boiling Point 993°C
Melting Point 620°C
Color Blue to Blue-Green
pH 3
Physical Form Solid
Quantity 100 g
Grade Pure
Packaging Glass Bottle
Specific Gravity 2.54
Chemical Name or Material Cupric Chloride Dihydrate, Purified (Copper (II) Chloride Dihydrate)