missing translation for 'onlineSavingsMsg'
Learn More

Cupric Chloride, Dihydrate, Crystal, ACS, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor C138525GM

 View more versions of this product

Catalog No. 18609991


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

10125-13-0
25 g
ACS
1
99%
Amber Glass Bottle

Chemical Identifiers

10125-13-0
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.