missing translation for 'onlineSavingsMsg'
Learn More

Cupric Chloride, Dihydrate, Crystal, ACS, 99%, Spectrum™ Chemical

Catalog No. 18609990
Change view
Click to view available options
Quantity:
5 g
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609990 5 g Amber Glass Bottle
18609991 25 g Amber Glass Bottle
18609992 125 g Amber Glass Bottle
18609993 500 g Amber Glass Bottle
18609994 2.5 kg Amber Glass Bottle
18609995 12 kg Poly Pail
18609996 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18609990 Supplier Spectrum Chemical Mfg Cor Supplier No. C13855GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 10125-13-0

Specifications

CAS Max % 1
Quantity 5 g
Percent Purity 99%
Grade ACS
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.