missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cupric Chloride Dihydrate, ACS Reagent (Copper (II) Chloride Dihydrate), Reagents
Cupric Chloride Dihydrate, ACS Reagent (Copper (II) Chloride Dihydrate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.
$228.50 - $1951.50
Chemical Identifiers
| Catalog Number | Mfr. No. | Price | Quantity | ||||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Price | Quantity | ||||||
|
01-338-038
|
Labchem Inc
C11885001000C2 |
Each for $1,951.50
|
|
||||||
|
01-338-037
|
Labchem Inc
C1188500100C2 |
Each for $228.50
|
|
||||||
|
01-338-036
|
Labchem Inc
C1188500500C2 |
Each for $983.68
|
|
||||||
Chemical Identifiers
| 0.0 |
Specifications
| 993°C | |
| Blue or Blue-Green | |
| 3 | |
| 1 kg | |
| MFCD00149674 | |
| MPTQRFCYZCXJFQ-UHFFFAOYSA-L | |
| copper(2+) dihydrate dichloride | |
| ACS | |
| 2.54 |
| 620°C | |
| 10125-13-0 | |
| Solid | |
| Cl2CuH4O2 | |
| Copper(II) Chloride Dihydrate | |
| O.O.[Cl-].[Cl-].[Cu++] | |
| 170.48 | |
| Glass Bottle | |
| Cupric Chloride Dihydrate, ACS Reagent (Copper (II) Chloride Dihydrate) |