Learn More
Cupferron, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 423605000
| Quantity | 500g |
|---|
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C6H9N3O2 | |
| MFCD00078422 | |
| cupferron, copperone, azanyliumylidene n-nitroso-n-oxidoaniline | |
| azanium;N-oxido-N-phenylnitrous amide |
Specifications
| Cupferron | |
| Plastic bottle | |
| C6H4N(ONH4)NO | |
| MFCD00078422 | |
| 15,2612 | |
| Solubility in water: decomposes. Other solubilities: soluble in alcohol,ether | |
| C1=CC=C(C=C1)N(N=O)[O-].[NH4+] | |
| 155.16 | |
| 155.16 |
| 135-20-6 | |
| C6H9N3O2 | |
| 500g | |
| 16,I,395 | |
| cupferron, copperone, azanyliumylidene n-nitroso-n-oxidoaniline | |
| GDEBSAWXIHEMNF-UHFFFAOYSA-O | |
| azanium;N-oxido-N-phenylnitrous amide | |
| 2724103 | |
| ACS Reagent |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Toxic if swallowed.
Suspected of causing cancer.
GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective glo
GHS Signal Word: Danger
EINECSNumber : 205-183-2
RTECSNumber : NC4725000
TSCA : TSCA
Promotions