missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Culture Tubes for SafTest™ Test Kits

Culture Tubes for SafTest™ Test Kits contains replacement consumables to run all SafTest test kits.

Supplier:  MP Biomedicals™ 07DP1041

Catalog No. MP07DP1041


Only null left
Explore available promotions

Description

Description

  • For use with SafTest analyzer
  • Used with all six SafTest test kits
  • Appropriate for all testing endpoints
  • Disposable
  • Packaged in shrink wrapped trays to prevent contamination
  • Size: 12 x 75 mm
  • Construction: Borosillicate glass
  • Used for tissue, cell cultures
Specifications

Specifications

75 mm
Single-use
Packaged in shrink wrapped trays to prevent contamination
Non-sterile
Borosilicate Glass
Used for tissue, cell cultures
250 Pk.
12 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions