Learn More
Thermo Scientific™ CultiMaxx™ Shelving System for G-Rex 500M-CS Bioreactors
CultiMaxx™ Shelving Systems to support G-Rex 500M-CS Bioreactors
$11,833.07
| Catalog Number | Mfr. No. | No. of Shelves | Material | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | No. of Shelves | Material | Price | Quantity & Availability | ||||
|
11670000
|
Thermo Scientific™
50164781 |
3 | Stainless Steel (18/8) |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Maximize the capacity, increase throughput and optimize cell growth in your lab using the CultiMaxx shelving system in a range of large capacity CO2 Incubators.
Maximize incubator space
- These shelving systems are designed to support scale-out of the G-Rex 500M-CS* in cell therapy production.
- With an in-depth review of the shelving design, this new system provides up to a 150% increase in capacity for the G-Rex, or another way to look at it: 2 1/2 incubators in 1.
Easy installation
- Shelving systems are retrofittable and can be self-installed.