Learn More
Thermo Scientific™ CultiMaxx™ Shelving Systems for Cell Culture Bags
Catalog No. p-200008780
Change view
Click to view available options
For Use With (Equipment):
Heracell VIOS 250i CO2 Incubator
No. of Shelves:
8
Catalog No. For Use With (Equipment) No. of Shelves
01257029 Heracell VIOS 250i CO2 Incubator 8
1 options
Catalog No. 01257029 Supplier Thermo Scientific™ Supplier No. 50166275
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CultiMaxx™ Shelving System to support Cell Culture Bags
Maximize the capacity, increase throughput and optimize cell growth in your lab using the CultiMaxx™ Shelving Systems in a range of large capacity CO2 Incubators.
These shelving systems are designed to support scale-out of cell culture bags in cell therapy production. With an in-depth review of the shelving design, this new system provides up to 3X more vessel capacity per unit. The shelving is specifically designed to promote greater gas exchange with increased exposure to the bottom of the semi-permeable bag. Since they are gas permeable on both sides, CultiMaxx shelving for cell culture bags will increase gas exchange compared to standard shelving. These shelves extend to support individual access and improved ease of handling.
Specifications
Description Shelving system to support Cell Culture Bags with base set for Heracell™ Vios™ 250i CO2 Incubator
For Use With (Equipment) Heracell VIOS 250i CO2 Incubator
Material Stainless Steel
No. of Shelves 8
Type Shelving System
Includes 8 shelves, supports and rails