missing translation for 'onlineSavingsMsg'
Learn More

Cucurbitacin B 98.0+%, TCI America™

Catalog No. C34425MG
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C34425MG 5 mg
C344225MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C34425MG Supplier TCI America Supplier No. C34425MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6199-67-3
Molecular Formula C32H46O8
Molecular Weight (g/mol) 558.712
MDL Number MFCD07778083
InChI Key IXQKXEUSCPEQRD-DKRGWESNSA-N
PubChem CID 5281316
ChEBI CHEBI:3941
IUPAC Name [(E,6R)-6-[(2S,8S,9R,10R,13R,14S,16R,17R)-2,16-dihydroxy-4,4,9,13,14-pentamethyl-3,11-dioxo-2,7,8,10,12,15,16,17-octahydro-1H-cyclopenta[a]phenanthren-17-yl]-6-hydroxy-2-methyl-5-oxohept-3-en-2-yl] acetate
SMILES CC(=O)OC(C)(C)C=CC(=O)C(C)(C1C(CC2(C1(CC(=O)C3(C2CC=C4C3CC(C(=O)C4(C)C)O)C)C)C)O)O

Specifications

Melting Point 179°C
Color White-Yellow
Quantity 5 mg
UN Number 2811
Formula Weight 558.71
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Cucurbitacin B
RTECSNumber RC6190000
TSCA No
Recommended Storage Refrigerator