missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Cucurbic Acid (5mg/mL in Acetonitrile), TCI America™

Catalog No. C20541ML
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20541ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C20541ML Supplier TCI America Supplier No. C20541ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 131488-83-0
Molecular Formula C12H20O3
Molecular Weight (g/mol) 212.289
InChI Key LYSGIJUGUGJIPS-PXLFNHBNSA-N
Synonym (+/-)-3-Hydroxy-2-(2-pentenyl)cyclopentaneacetic Acid
PubChem CID 131872090
IUPAC Name 2-[(1S,2R,3R)-3-hydroxy-2-[(Z)-pent-2-enyl]cyclopentyl]acetic acid
SMILES CCC=CCC1C(CCC1O)CC(=O)O

Specifications

Quantity 1 mL
UN Number 1648
Formula Weight 212.29
Chemical Name or Material (+/-)-Cucurbic Acid (5mg/mL in Acetonitrile)