missing translation for 'onlineSavingsMsg'
Learn More

TCI America™ CUBIC-HV™ 3D Immunostaining Kit (Casein Separate)

Catalog No. C37171KIT
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C37171KIT 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C37171KIT Supplier TCI America™ Supplier No. C37171KIT
Only null left

This kit contains buffers and anti-NeuN antibody (positive control) for 3D immunostaining of CUBIC-cleared tissues. It is suitable for the use on the brain of an adult mouse. Please note that CUBIC-L (T3740) and CUBIC-R+(M) (T3741) are required to clear tissue first.

This kit contains the following:

  • CUBIC-HV™1 3D immunostaining buffer (without Casein) (2x) (for 10 tests)
  • CUBIC-HV™1 3D immunostaining wash buffer (1x) (for 10 tests)
  • CUBIC-HV™1 3D immunostaining additive (10x) (for 10 tests)
  • Anti-NeuN Mouse IgG1 antibody (1 mg/mL) (for 2 tests)
  • 10 x 15 mL tube

Note: The difference with this product and C3708-KIT is that this product includes a buffer without casein and casein is in the kit separately. The casein should be mixed into the buffer just before use and, after mixing, the buffer should be used as soon as possible. It takes about 2 hours to solve the casein.

Specifications

For Use With (Equipment) Light-sheet Fluorescent Microscopy (LSFM) or Confocal Laser-scanning Microscopy (CLSM) Device
Product Line CUBIC
Quantity 1 Kit
For Use With (Application) Neuroscience, 3D pathology, oncology, histology, immunology, and other research applications
Form Liquid