missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Cu/Zn Superoxide Dismutase ELISA Kit
Description
Includes
Pre-Coated 96-Well Plate, Anti-Cu/Zn SOD/HRP-Conjugate, Cu/Zn SOD Standard, Wash Buffer Concentrate, Assay Buffer Concentrate, Phosphate Buffered Saline, Substrate Solution, Blue and Green Dyes, Stop Solution, Plate Covers, and a user protocol
A colorimetric sandwich ELISA method for detecting Cu/Zn superoxide dismutase (SOD) in different types of biological samples
Specifications
Specifications
| Content And Storage | store the entire contents of the kit at 4°C |
| Detection Method | Colorimetric |
| For Use With (Application) | ELISA |
| Quantity | 1 Kit |