missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ Xenon™ Lower Conductivity Electroporation Buffer

Catalog No. p-200019994
Change view
Click to view available options
Format:
Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
14259309 Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14259309 Supplier Gibco™ Supplier No. A5788001

CTS Xenon Lower Conductivity Electroporation Buffer is designed specifically for use with the Xenon Electroporation System.

Gibco CTS Xenon Lower Conductivity Electroporation Buffer is designed specifically for use with the Xenon Electroporation System. It is recommended for use with cell types that require higher energy electroporation settings to achieve successful transfection. The formulation helps protect against excessive heat generation during the electroporation process.

Closed-system compatibility

CTS Xenon Lower Conductivity Electroporation Buffer is available in 100-mL bottle and 100-mL bag formats for convenient process development and closed-system manufacturing scale-up.

CTS quality

Gibco CTS products follow USP 'ancillary materials for cell, gene, and tissue-engineered products' within the responsibilities applicable to a supplier.* The products are supplied with harmonized documentation such as intended use statements, certificates of analysis, certificates of origin, and convenient access to our drug master file (DMF). Full documentation traceability is available to support your regulatory filing.

Gibco CTS products are manufactured at a site that uses methods and controls that conform to cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485-certified.

*Other aspects of USP will be the responsibility of the end user to assess. Thermo Fisher Scientific cannot fulfill USP in regard to application and therapy-specific aspects (e.g., residual assessment and removal of the AM).

Specifications

Format Bottle
For Use With (Equipment) CTS™ Xenon™ Electroporation System
For Use With (Application) Cell and Gene Therapy Research, Development and Manufacturing
Product Type Electroporation Buffer
Quantity 100 mL bottle
Shipping Condition Gel packs
Volume (Metric) 100 mL
Manufacturing Quality cGMP for Medical Devices, 21 CFR Part 820, and ISO 13485

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.