missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ Viral-Plex™ Complexation Buffer

Catalog No. A5145403
Change view
Click to view available options
Quantity:
500 mL
1 L
10 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5145403 10 L
A5145401 500 mL
A5145402 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A5145403 Supplier Gibco™ Supplier No. A5145403

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Gibco CTS Viral-Plex Complexation Buffer is a chemically defined, protein-free, phenol-red-free buffer for complexing plasmid DNA with a transfection reagent for standard transfection applications.

Gibco CTS Viral-Plex Complexation Buffer is a chemically defined, protein-free, phenol-red-free buffer for complexing plasmid DNA with a transfection reagent for standard transfection applications. It is a part of the CTS AAV-MAX system and is designed to be used with the CTS AAV-MAX Transfection Kit for the transient transfection of CTS Viral Production Cells 2.0.

This buffer is manufactured in conformity with cGMP for medical devices (21 CFR Part 820) and follows USP <1043> and European Pharmacopoeia (Ph. Eur.) 5.2.12 recommendations. A Drug Master File and Regulatory Support File are available upon request.

The CTS AAV-MAX Helper-Free AAV Production System is a scalable, high-titer platform for suspension AAV production with research-grade (Cat. No. A51217) and GMP versions to enable seamless transition from research to commercial manufacturing. Other components of the CTS AAV-MAX system include CTS Viral Production Cells 2.0 (A48400), CTS Viral Production Medium (A5144001), CTS AAV-MAX Transfection Kit (A5427701), and Viral Production Medium, AGT (A5147101).

Specifications

For Use With (Application) Transient transfection for mammalian AAV production
Physical Form Liquid
Quantity 10 L
Packaging BPC
Sterility Sterile-filtered
Manufacturing Quality ISO 13485, MDSAP, FDA-registered, 21 CFR 820