missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ StemPro™-34 SFM XF Medium

Catalog No. A6636901
Change view
Click to view available options
Quantity:
500 mL
2 L
Product Type:
Bottle Kit
BPC Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
A6636901 500 mL Bottle Kit
A6636902 2 L BPC Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A6636901 Supplier Gibco™ Supplier No. A6636901

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Gibco CTS StemPro-34 SF XenoFree Medium is a flexible, optimized medium for hematopoietic stem and progenitor cell applications.

Gibco CTS StemPro-34 SF XenoFree Medium is a highly effective medium designed for the growth and development of human hematopoietic cells in culture, including hematopoietic stem cells (HSCs) and progenitor cells sourced from bone marrow, neonatal cord blood, and peripheral blood.

Key benefits for cell and gene therapy applications

  • Serum-free and xeno-free—formulated without serum or non-human derived components to comply with global regulatory standards for ancillary materials in cell therapy manufacturing; formulated with L-glutamine and without phenol red
  • Supports cell differentiation—facilitates differentiation of induced pluripotent stem cells (iPSCs) to HSCs and further differentiation into immune cells (NK cells, T-cells, B-cells, macrophages, etc.)

Regulatory compliance and quality assurance

  • cGMP manufactured—produced in accordance with 21 CFR part 820 and ISO 9001 and ISO 13485 standards to help ensure the highest quality and safety
  • USP <1043> compliant—designed to meet USP <1043> standards for ancillary materials used in cell, gene, and tissue-engineered products

Seamless integration with closed system workflows

CTS StemPro-34 SF XenoFree Medium is available in both 500-mL bottle and 2-liter bioprocessing container (BPC) formats, supporting processes through cell therapy development and manufacturing. The BPC chamber is constructed from Aegis 5-14 film and includes sterile, weldable DEHP-free PVC tubing with Luer lock and C-Flex tubing with MPC quick connector. This helps provide versatile connection options, helping ensure maximum sterility and efficiency in your manufacturing process. The 2-L kit format includes both supplement and basal components in BPCs for closed system workflows.

Comprehensive documentation for regulatory support

To facilitate your regulatory submissions, CTS StemPro-34 SF XenoFree Medium comes with an extensive documentation package, including certificates of analysis (COA), access to drug master files (DMFs), and/or regulatory support files (RSFs), and certificates of origin (COO) upon request.

Choose CTS StemPro-34 SF XenoFree Medium for a robust, regulatory-compliant solution that supports the critical processes in cell and gene therapy manufacturing, helping ensure the highest standards of quality and safety.

Specifications

Classification Xeno-free, Serum-free, cGMP
Form Liquid
Product Type Bottle Kit
Serum Level Serum-free
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives Glutamine
Without Additives No Phenol Red
Quantity 500 mL
Culture Type Stem Cell (Human, iPS - Induced Pluripotent Stem, Embryonic), Suspension Cell Culture, 3D Cell Culture
What is CTS?

The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.

Once thawed, can CTS StemPro-34 Supplement in bags be refrozen?

Once thawed, CTS StemPro-34 Supplement should not be refrozen in the bag. If necessary, the supplement can be removed from the bag to a secondary container and refrozen. Avoid additional freeze/thaw cycles.

How long can complete CTS StemPro-34 SFM XF medium (basal medium plus supplement) be stored at 4 degrees C?

Complete CTS StemPro-34 SFM XF medium can be stored for 4 weeks at 4 degrees C, however, we recommend adding growth factors or cytokines just prior to use.

How do I thaw and mix CTS StemPro-34 Supplement?

CTS StemPro-34 Supplement can be thawed at room temperature (RT) for ~1 hour or at 4 degrees C overnight. Do not thaw the frozen supplement at 37 degrees C. Once completely thawed, the supplement should be allowed to equilibrate to RT for at least 10 min, but not longer than 3 hr, before gently mixing the supplement by inverting the vial 6-8 times until the supplement is completely homogenous.

How should I store the components of the CTS StemPro-34 SFM XF kit?

CTS StemPro-34 Basal Medium should be stored at 2 to 8 degrees C, protected from light. CTS StemPro-34 Supplement should be stored at -20 to -5 degrees C, protected from light.

Does CTS StemPro-34 SFM XF contain any added growth factors or cytokines?

CTS StemPro‑34 SFM XF is manufactured without cytokines or growth factors, to allow the researcher flexibility for the intended application. Please refer to the user guide for details.

What types of cells have been tested with CTS StemPro-34 SFM XF?

CTS StemPro-34 SFM XF can support growth of human hematopoietic stem and progenitor cells in culture, including CD34+ cells from bone marrow, neonatal cord blood, and peripheral blood, when supplemented with appropriate growth factors or cytokines. In addition, CTS StemPro-34 SFM XF can support differentiation of human induced pluripotent stem cells (iPSCs) to iPSC-derived HSPCs, when supplemented with appropriate growth factors, cytokines and other additions. A suggested protocol is available here.

What are the main differences between StemPro-34 SFM and CTS StemPro-34 SFM XF?

CTS Stempro-34 SFM XF is for research use or manufacturing of cell-, gene-, or tissue-based products. It is not intended for direct administration into humans or animals. It is serum-free and xeno-free, contains L-Glutamine and does not contain phenol red. For a detailed description of CTS StemPro-34 SFM XF, please refer to the Product Overview section on the product page.

StemPro-34 SFM is designed for research use only, it is serum-free, contains phenol red, and requires addition of a source of L-Glutamine.


For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.