missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ OpTmizer™ T Cell Expansion SFM, bottle format

Catalog No. a1048501
Encompass
Change view
Click to view available options
Quantity:
1000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1048501 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1048501 Supplier Gibco™ Supplier No. A1048501
Only null left

CTS™ OpTmizer™ T Cell Expansion SFM, bottle format

GIBCO OpTmizer™ CTS™ T-Cell Expansion SFM has been developed for the growth and expansion of human T lymphocytes. OpTmizer™ CTS™ T-Cell Expansion medium is a complete serum-free, xeno-free 1X medium consisting of OpTmizer™ T-Cell Expansion Basal Medium (1 L bottle) and OpTmizer™ T-Cell Expansion Supplement (26 mL), which are mixed together prior to use.

  • Maintains similar phenotype and function (e.g., cytokine secretion profile) of polyclonally activated and cultured T cells as observed for conventional serum supplemented medium
  • Consistent performance in supporting T cell expansion
  • Supports high density T cell culture (e.g., >3x106 CD3+ T cells/mL) in static culture

    With OpTmizer™ T-Cell Expansion SFM, we offer both basic and clinical research communities a new tool to enhance their success in growing human T-cells. It is specially formulated to allow for superior cell growth & viability, less variability and more consistent results, and easier transition from research to clinical applications.

    Workflow benefits:
  • Minimizing the need to handle purchasing, storage, qualification of serum and thus reducing variability and costs
  • More rapid T cell expansion, thus reducing cycle time and improving productivity
  • Complete serum-free, xeno-free medium, allowing for a seamless transition from research to clinical applications and minimizing regulatory workload

    “This reagent is classified as a medical device in several countries. If it is not regulated as a medical device in specific region, it should be used for laboratory use only.”
  • Specifications

    Cell Type T Cells
    Classification Serum-free, Xeno-free
    Form Liquid
    Product Type Cell Expansion Serum Free Medium (SFM)
    Serum Level Serum-free
    Sterility Sterile
    With Additives HEPES, Phenol Red, Sodium Pyruvate, Sodium Bicarbonate
    Without Additives No Glutamine
    Product Line CTS, OpTmizer
    Quantity 1000 mL
    Endotoxin Level Very Low
    Included Antibiotics No antibiotics
    Inorganic Salts Calcium, Magnesium
    Purity or Quality Grade Research Grade
    Content And Storage OpTmizer™ T-Cell Expansion Basal Medium (1 x 1000 mL bottle): Store at 2–8°C. Protect from light.

    OpTmizer™ T-Cell Expansion Supplement (1 x 26 mL): Store in the dark at 2–8°C.
    Culture Environment CO2
    Culture Type Suspension Cell Culture
    Show More Show Less