missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ OpTmizer™ Pro SFM, bottle format

Catalog No. A4966101
Change view
Click to view available options
Quantity:
1000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A4966101 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A4966101 Supplier Gibco™ Supplier No. A4966101

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Gibco CTS OpTmizer Pro SFM, no phenol red, is a novel serum-free medium (SFM) developed for the growth and expansion of human T lymphocytes (e.g., CD4+, CD8+, polyclonal, or antigen specific) used in allogeneic cell therapies.

Gibco CTS OpTmizer Pro SFM, no phenol red, is a novel serum-free medium (SFM) developed for the growth and expansion of human T lymphocytes (e.g., CD4+, CD8+, polyclonal, or antigen specific) used in allogeneic cell therapies. This medium improves central memory phenotype and cell growth by shifting the cell's metabolism. The result is a larger population of central memory cells in a shorter period of time.

Benefits

  • Maintains central memory T cells further into expansion and is well suited for longer workflows (>15 days)
  • Offers >90% (average) T cell fold expansion (from healthy donors) compared to CTS OpTmizer medium
  • Modest increases in central memory cells at early stage in expansion protocol result in larger cell yields at harvest
  • Helps achieve the same number of cells in a shorter duration
  • Ideally suited to the production of allogeneic therapies

Features

  • Serum-free and xeno-free
  • Supports high density cultures in static cultures
  • New film (Aegis) and bag design (Cat. No. A4966101) allow compatibility with closed systems
  • Supports T-cell activation using Dynabeads magnetic beads, soluble antibodies, and stimulatory antibody-presenting cell protocols

Increased efficiency and consistence performance

CTS OpTmizer Pro SFM, no phenol red, is serum-free and xeno-free and supports high density cultures in static cultures. It displays consistent performance in the support of T-cell expansion with better central memory phenotype in a shorter time.

Enhanced expansion

CTS OpTmizer Pro SFM, no phenol red, increases the number of central memory cells at an early stage in the expansion process and maintains T-cell functionality.

Simplify your transition into clinical research

The Gibco Cell Therapy Systems (CTS) brand offers a broad array of products for use in cell therapy applications, including media, reagents, growth factors, enzymes, cell selection/expansion reagents, and devices. CTS products deliver:

  • Reduced burden in qualifying reagents
  • Appropriate certificates of analysis and origin available
  • Convenient access to our Drug Master File (DMF)

Specifications

Cell Type T Cells
Classification Xeno-free
Form Liquid
Product Type Cell Culture Medium
Serum Level Serum-free
Sterility Sterile
Without Additives No Glutamine, No Phenol Red
For Use With (Application) Cell Therapy, CAR-T, Allogeneic, T Cell Expansion Media, Cancer Immunotherapy
Quantity 1000 mL
Shipping Condition Room Temp
Tested For Endotoxin
Culture Type Suspension Cell Culture
Show More Show Less