Learn More
Gibco™ CTS™ OpTmizer™ One SFM
Description
Gibco CTS OpTmizer One SFM is a novel animal-origin-free (AOF) serum-free medium (SFM) formulation developed for the growth and expansion of human T lymphocytes used in cell therapies. This medium improves cell count and early memory phenotype throughout all stages of expansion.
Benefits:
- Provides enhanced T-cell expansion and expression of early memory phenotype makers compared to other chemically defined media
- Demonstrates flexibility across culture vessels. In both G-Rex™ plates and rocking bioreactors, it supports scale-up while maintaining desired T-cell phenotype.
- Expands both healthy and disease state donors
- Supports efficient viral and non-viral editing with downstream expansion
- Mitigates risk of contaminants and variability associated with animal-sourced materials
- Supports scale-up via larger catalog pack sizes
Features:
- Animal-origin-free (AOF) formulation
- Serum-free—designed without the need for serum or serum replacement
- No separate components—no need to mix basal medium with supplement to obtain complete medium
- Yields high density cultures
- Supports T-cell activation using CTS Dynabeads (both passive release CD3/CD28 (Cat. No.40203D) and detachable CD3/CD28 (Cat No. A56992))
- Available in 1 L bottle, 1 L bag, 5 L bag, and 10 L bag options
Increased efficiency and consistent performance:
CTS OpTmizer One SFM is an AOF formulation that supports high density cultures in static and dynamic vessels. It displays consistent performance for T-cell expansion, with better early memory phenotype at early stages.
Simplify your transition into clinical research:
The Gibco Cell Therapy Systems (CTS) portfolio offers a broad array of products for use in cell therapy applications, including media, reagents, growth factors, enzymes, cell selection/expansion reagents, and devices. CTS products deliver:
- Reduced burden in qualifying reagents
- Appropriate certificates of analysis and origin available
- Convenient access to our Drug Master File (DMF)
Specifications
Specifications
| Cell Type | T cells |
| Classification | Animal Origin-free, Serum-free, cGMP |
| Form | Liquid |
| Serum Level | Serum-free |
| Sterility | Sterile-filtered |
| Sterilization Method | 0.2 μm Sterile-filtered |
| With Additives | None |
| Without Additives | No Glutamine, No Phenol Red |
| Product Line | CTS OpTmizer |
| Quantity | 5 L |
| Show More |
For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.