Learn More
Gibco™ CTS™ Opti-MEM™ I Medium
Description
Gibco™ CTS Opti-MEM I Medium is an improved minimal essential medium (MEM) that is recommended for use with the CTS LV-MAX Transfection Kit as a complexation buffer. This medium contains insulin, transferrin, hypoxanthine, thymidine, and trace elements. CTS Opti-MEM I uses a sodium bicarbonate buffer system (2.4 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH. The formulation of CTS Opti-MEM I is identical to research-grade Opti-MEM I Reduced Serum Medium.
CTS Opti-MEM I is xeno-free and contains human transferrin.
CTS Opti-MEM I is manufactured in conformity with cGMP for medical devices, 21 CFR, Part 820, and certified to the ISO 13485. Drug Master File and Regulatory Support File available upon request.
Custom configuration and packaging available upon request.
Spécifications
Spécifications
| Lignée cellulaire | Cellules de production virale |
| Type de cellules | HEK293 |
| Classification | Sans origine animale |
| Forme | Liquide |
| Type de produit | Médium essentiel minimum (MEM) |
| Stérilité | Filtré stérile |
| À utiliser avec (application) | LV Production |
| Qualité de fabrication | ISO 13485, MDSAP, enregistré auprès de la FDA, 21 CFR 820 |
| Quantité | 100 mL |
| Shipping condition (Condition d'expédition) | Conditions ambiantes |
| Afficher plus de résultats |
Pour la recherche ou la fabrication de produits à base de cellules, de gènes ou de tissus. ATTENTION : Pas destiné à être administré directement aux humains ou aux animaux.