missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ Neurobasal™-A Medium
GREENER_CHOICE

Catalog No. a1371001
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1371001 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1371001 Supplier Gibco™ Supplier No. A1371001
Only null left

CTS Neurobasal-A Medium is designed with the translational stem cell researcher in mind. It can be used to culture post-natal neurons without the need for an astrocyte feeder layer when used with B-27 supplements.

CTS Neurobasal-A Medium is designed with the translational stem cell researcher in mind. It can be used to culture post-natal neurons without the need for an astrocyte feeder layer when used with B-27 supplements.

CTS quality

Gibco CTS products are manufactured at a site that uses methods and controls that conform with cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485– and ISO 9001–certified.

Accompanied by documentation to support your regulatory filing

Specific intended-use statements, full documentation traceability, and convenient access to our Drug Master File (DMF) are available.

For other applications, we provide alternative formulations of CTS Neurobasal media and B-27 supplements.

For established protocols using Neurobasal media, access our Gibco Neurobiology Protocol Handbook.

View publications using Gibco neurobiology products at our Gibco CellCite.

Specifications

Cell Type Neural Cells
Classification Animal Origin-free
Form Liquid
Product Type Cell Culture Medium
Serum Level Serum-free
Sterility Sterile-filtered
For Use With (Application) Cell and Gene Therapy Research, Development, and Manufacturing
Manufacturing Quality ISO 13485, MDSAP, FDA-registered, 21 CFR 820
Quantity 500 mL
Shipping Condition Ambient
Format Bottle
Content And Storage Store at 2–8°C and protect from light.
Show More Show Less

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.