missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ KnockOut™ SR XenoFree Medium
GREENER_CHOICE

Catalog No. p-7075762
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12618013 500 mL
12618012 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12618013 Supplier Gibco™ Supplier No. 12618013
Only null left

CTS KnockOut SR XenoFree Medium enables the growth and expansion of human embryonic stem cells (hESCs) and human induced pluripotent stem cells (hiPSCs) in a cell culture medium containing only human-derived or human recombinant proteins.

Gibco CTS KnockOut SR XenoFree Medium, when formulated as a complete medium, enables the growth and expansion of human embryonic stem cells (hESCs) and human induced pluripotent stem cells (hiPSCs) in a cell culture medium containing only human-derived or human recombinant proteins, to facilitate the transition of hESC and hiPSC research from the bench to the clinic.

Reduces risks

CTS KnockOut SR XenoFree Medium does not contain bovine or other non-human, animal-derived components.

Supports multiple applications

Besides pluripotent stem cell culture expansion and maintenance, CTS KnockOut SR XenoFree Medium can be used for hESC/hiPSC cryopreservation, derivation, and differentiation studies.

Facilitates regulatory filings

The CTS product line enables you to reduce your burden in qualifying reagents during your transition from research applications to clinical applications. Gibco CTS products are of high quality and are supplied with harmonized documentation such as Certificates of Analysis, Certificates of Origin, and access to authorization letters for our Drug Master Files, as appropriate.

Specifications

Cell Type Embryonic Stem Cells (ESCs), Induced Pluripotent Stem Cells (iPSCs)
Classification Xeno-free
Form Liquid
Product Type Cell Culture Supplement
Serum Level Serum-free
Sterility Sterile-filtered
For Use With (Application) Cell and Gene Therapy Research, Development, and Manufacturing
Manufacturing Quality ISO 13485, MDSAP, FDA-registered, 21 CFR 820
Quantity 500 mL
Shipping Condition Dry Ice
Format Bottle
Content And Storage Store at –5 to –20°C and protect from light.
Show More Show Less

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.