Learn More
Gibco™ CTS™ KnockOut™ SR XenoFree Medium

Description
Gibco CTS KnockOut SR XenoFree Medium, when formulated as a complete medium, enables the growth and expansion of human embryonic stem cells (hESCs) and human induced pluripotent stem cells (hiPSCs) in a cell culture medium containing only human-derived or human recombinant proteins, to facilitate the transition of hESC and hiPSC research from the bench to the clinic.
Reduces risks
CTS KnockOut SR XenoFree Medium does not contain bovine or other non-human, animal-derived components.
Supports multiple applications
Besides pluripotent stem cell culture expansion and maintenance, CTS KnockOut SR XenoFree Medium can be used for hESC/hiPSC cryopreservation, derivation, and differentiation studies.
Facilitates regulatory filings
The CTS product line enables you to reduce your burden in qualifying reagents during your transition from research applications to clinical applications. Gibco CTS products are of high quality and are supplied with harmonized documentation such as Certificates of Analysis, Certificates of Origin, and access to authorization letters for our Drug Master Files, as appropriate.
Specifications
Specifications
| Cell Type | Embryonic Stem Cells (ESCs), Induced Pluripotent Stem Cells (iPSCs) |
| Classification | Xeno-free |
| Form | Liquid |
| Product Type | Cell Culture Supplement |
| Serum Level | Serum-free |
| Sterility | Sterile-filtered |
| For Use With (Application) | Cell and Gene Therapy Research, Development, and Manufacturing |
| Manufacturing Quality | ISO 13485, MDSAP, FDA-registered, 21 CFR 820 |
| Quantity | 500 mL |
| Shipping Condition | Dry Ice |
| Show More |
Frequently Asked Questions (FAQs)
The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.
Yes, we do offer a Class II medical device–labeled CTS KnockOut SR XenoFree Medium in the USA and Canada only. Please contact us at custommedia@thermofisher.com to discuss if a medical device grade is required for your application.
Human ES cells are generally characterized by their typical morphology (they grow as tightly packed clusters of small cells with high ratio of nucleus to cytoplasm); surface marker expression; RT-PCR detection of stem cell-specific gene expression (such as Oct3/4, Sox2, and Nanog); alkaline phosphatase staining, and telomerase activity assay. The most commonly used ES specific surface markers include stage-specific embryonic antigens SSEA-3 and SSEA-4 for human ES cells. Other ES-specific surface antigens also include TRA-1-60 and TRA-1-81. (Science 282:1145 (1998).
Human ES cells are derived from human blastocyst inner cell masses, isolated by immunosurgery with rabbit antiserum to BeWO cells (a human trophoblast cell line) (Science 282:1145 (1998)).
Embryonic stem (ES) cells are derived from the early mammalian embryo and are capable of unlimited, undifferentiated proliferation in vitro while maintaining their potential to differentiate into a wide of range of adult tissues including germ cells. The pluripotency of the ES cells is normally demonstrated in vitro by inducing ES cells to differentiate into embryoid bodies and checking lineage-specific markers for differentiated cells in three body layers (endo, meso, and ectoderm), or injecting them into immunodeficient mice and determining the cell types produced in the teratomas.
For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.