Learn More
Gibco™ CTS™ Essential 6™ Medium
Description
CTS Essential 6 Medium is a cGMP-manufactured, xeno-free, feeder-free medium that supports the reprogramming of somatic cells and the spontaneous or directed differentiation of human pluripotent stem cells (PSCs). The formulation is based on a medium originally developed by Guokai Chen, et al., in the laboratory of James Thomson (1). With only six essential components, CTS Essential 6 Medium helps minimize variability. To complete your PSC workflow with a matching culture medium for the growth and expansion of PSCs, use CTS Essential 8 Medium.
CTS Essential 6 Medium:
- Provides a flexible format to suit different applications
- Facilitates regulatory filings through cGMP manufacturing and the availability of documentation
- Simplifies the transition from bench to clinic through the direct replacement of RUO Essential 6 Medium
Provides a flexible format
CTS Essential 6 Medium suits different applications and various cell types. It does not contain bFGF or TGFβ and can therefore support the formation of embryoid bodies. This medium can support the directed differentiation of various cell types and somatic cell reprogramming using a variety of methods.
Facilitates regulatory filings
CTS Essential 6 Medium is manufactured at a site that uses methods and controls that conform with cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing site is ISO 13485-certified. Full documentation traceability and convenient access to our Drug Master File (DMF) are available for CTS Essential 6 Medium.
Simplifies transition from bench to clinic
CTS Essential 6 Medium contains the same six essential components as RUO Essential 6 Medium and, with its cGMP manufacturing process, this fully-defined formulation helps minimize variability. Like RUO Essential 6 Medium, CTS Essential 6 Medium does not contain bFGF or TGF-beta, which inhibit differentiation and negatively affect reprogramming efficiency, respectively. With its additional safety testing, including adventitious agents, CTS Essential 6 Medium enables a smooth transition from RUO Essential 6 Medium.
Commercialized in partnership with Cellular Dynamics International.
Reference
1. Chen G, Gulbranson DR, Hou Z, Bolin JM, Ruotti V, Probasco MD, Smuga-Otto K, Howden SE, Diol NR, Propson NE, Wagner R, Lee GO, Antosiewicz-Bourget J, Teng JM, Thomson JA. Chemically defined conditions for human iPSC derivation and culture. Nat Methods. 2011 8(5):424-9.
Specifications
Specifications
| Classification | Xeno-free |
| Form | Liquid |
| Product Type | Cell Culture Medium |
| Sterility | Sterile-filtered |
| For Use With (Application) | Cell and Gene Therapy Research, Development, and Manufacturing |
| Manufacturing Quality | ISO 13485, MDSAP, FDA-registered, 21 CFR 820 |
| Quantity | 500 mL |
| Shipping Condition | Ambient |
| Format | Bottle |
| Content And Storage | Store at 2–8°C and protect from light. |
Frequently Asked Questions (FAQs)
The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.
For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.