missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ DynaCellect™ Cell Isolation Kit

Catalog No. A52300
Change view
Click to view available options
Quantity:
5 kits
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A52300 5 kits
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A52300 Supplier Gibco™ Supplier No. A52300

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Gibco CTS™ DynaCellect™ Cell Isolation Kit is specifically designed for the isolation and activation of immune cells in cell processing applications. The design consists of a 1 L isolation chamber, 8 PVC lines for the welding of input and output bags, a cassette, and tube organizer.

The CTS DynaCellect Cell Isolation Kit is a single-use, fit-for-purpose kit designed specifically for use with the CTS DynaCellect Magnetic Separation System, which is an automated instrument for T-cell isolation and activation, as well as magnetic bead removal.

Characteristics of the CTS DynaCellect Cell Isolation Kit include:
• Weldable tube material: 8 x DEHP-free PVC
• Working volume size: 10 mL to 1 L
• Bag chamber size: 1 L

This kit and the CTS DynaCellect Bead Removal Kit enable a range of cell processing applications such as isolation, depletion, and bead removal. They offer flexibility for process development and can optimized to support commercial scale manufacture.

Each kit has input/output sterile weldable tubes for flexible setup configurations. They are compatible with sterile welding and are gamma sterilized.

Specifications

Product Type Isolation Kit
Capacity (Metric) 1 L
Quantity 5 kits

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.