missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ Detachable Dynabeads™ Release Buffer

The Gibco CTS Detachable Dynabeads Release Buffer is for use with CTS Detachable Dynabeads magnetic beads. When used together, they empower users with a first-of-its-kind cGMP cell selection technology that has an active release mechanism for process development, clinical trial, and commercial manufacturing use.

$588.26 - $1,665.06

Spécifications

Color Clear
Concentration 5 mM
For Use With (Application) Cell Therapy
For Use With (Equipment) CTS DynaCellect Magnetic Separation System
pH 7.0 to 7.4
voir les spécifications

Produits 3
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantity & Availability  
A5588303
Afficher les documents Détails de la promotion
Gibco™
A5588303
212 mL
chaque for $588.26
Il en reste null
 
A5588301
Afficher les documents Détails de la promotion
Gibco™
A5588301
400 mL
chaque for $943.46
Il en reste null
 
A5588302
Afficher les documents Détails de la promotion
Gibco™
A5588302
750 mL
chaque for $1,665.06
Il en reste null
 
Description

Description

The Gibco CTS Detachable Dynabeads Release Buffer is for use with CTS Detachable Dynabeads magnetic beads. When used together, they empower users with a first-of-its-kind cGMP cell selection technology that has an active release mechanism for process development, clinical trial, and commercial manufacturing use.

Features of the CTS Detachable Dynabeads Release Buffer (when used with CTS Detachable Dynabeads magnetic beads) include:

  • Empowers users with process flexibility by allowing active release of beads from target cells at any time
  • Optimizes target cell purity, yield, activation profile, and desired phenotype thanks to the active release mechanism that gives users more control
  • Provides an automation-friendly solution by working directly with the CTS DynaCellect Magnetic Separation System
  • Enables consistent performance in light of biological variability within processes
  • Is suitable for both autologous and allogeneic cell therapy manufacturing processes

The CTS Detachable Dynabeads Release Buffer has an active ingredient, biotin, that competitively displaces a biotin derivative on the surface of CTS Detachable Dynabeads magnetic beads. This proprietary mechanism allows users to achieve active release of the beads from target cells at any time within their process. And when used on the CTS DynaCellect Magnetic Separation System, these products deliver automation, flexibility, and scalability in cell therapy manufacturing workflows.

The CTS Detachable Dynabeads Release Buffer also includes Drug Master Files (DMFs) to support cross-referencing in regulatory submissions.

Spécifications

Spécifications

Clear
Cell Therapy
7.0 to 7.4
Buffer
5 mM
CTS DynaCellect Magnetic Separation System
Liquid
GMP
Videos
Sécurité et manipulation

Sécurité et manipulation

Recommended Storage : Store at 2°C to 8°C and protect from light

FDS
Documents

Documents

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.