missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ Detachable Dynabeads™ Release Buffer

Catalog No. A5588303
Change view
Click to view available options
Quantity:
212 mL
400 mL
750 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5588303 212 mL
A5588301 400 mL
A5588302 750 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A5588303 Supplier Gibco™ Supplier No. A5588303

The Gibco CTS Detachable Dynabeads Release Buffer is for use with CTS Detachable Dynabeads magnetic beads. When used together, they empower users with a first-of-its-kind cGMP cell selection technology that has an active release mechanism for process development, clinical trial, and commercial manufacturing use.

The Gibco CTS Detachable Dynabeads Release Buffer is for use with CTS Detachable Dynabeads magnetic beads. When used together, they empower users with a first-of-its-kind cGMP cell selection technology that has an active release mechanism for process development, clinical trial, and commercial manufacturing use.

Features of the CTS Detachable Dynabeads Release Buffer (when used with CTS Detachable Dynabeads magnetic beads) include:

  • Empowers users with process flexibility by allowing active release of beads from target cells at any time
  • Optimizes target cell purity, yield, activation profile, and desired phenotype thanks to the active release mechanism that gives users more control
  • Provides an automation-friendly solution by working directly with the CTS DynaCellect Magnetic Separation System
  • Enables consistent performance in light of biological variability within processes
  • Is suitable for both autologous and allogeneic cell therapy manufacturing processes

The CTS Detachable Dynabeads Release Buffer has an active ingredient, biotin, that competitively displaces a biotin derivative on the surface of CTS Detachable Dynabeads magnetic beads. This proprietary mechanism allows users to achieve active release of the beads from target cells at any time within their process. And when used on the CTS DynaCellect Magnetic Separation System, these products deliver automation, flexibility, and scalability in cell therapy manufacturing workflows.

The CTS Detachable Dynabeads Release Buffer also includes Drug Master Files (DMFs) to support cross-referencing in regulatory submissions.

Specifications

Color Clear
Concentration 5 mM
For Use With (Application) Cell Therapy
For Use With (Equipment) CTS DynaCellect Magnetic Separation System
pH 7.0 to 7.4
Physical Form Liquid
Quantity 212 mL
Solution Type Buffer
Grade GMP

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.