missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ Detachable Dynabeads™ CD8

Catalog No. A56995
Change view
Click to view available options
Quantity:
15 mL vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A56995 15 mL vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A56995 Supplier Gibco™ Supplier No. A56995

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CTS Detachable Dynabeads CD8 for positive isolation of CD8+ T cells and active release of Dynabeads post-isolation.

Gibco CTS Detachable Dynabeads CD8 are intended for ex vivo isolation of CD8+ T cell subsets. Combined with CTS Detachable Dynabeads Release Buffer (Cat. No. A5588301, A5588302, or A5588303), it is a first-of-its-kind cGMP cell selection technology with an active release mechanism for process development, clinical trial, and commercial manufacturing use. This product is also commonly used with CTS Detachable Dynabeads CD4 (Cat. No. A56994).

Features of CTS Detachable Dynabeads CD8 include:

  • Empower users with process flexibility by allowing active release of beads from target cells at any time
  • Optimize target cell purity and yield thanks to the active release mechanism that gives users more control
  • Provide an automation-friendly solution by working directly with the CTS DynaCellect Magnetic Separation System
  • Enable consistent performance in light of biological variability within the starting material
  • Suitable for both autologous and allogeneic cell therapy manufacturing processes

These Detachable Dynabeads are meant for direct use with CTS Detachable Dynabeads Release Buffer, which has an active ingredient, biotin, that competitively displaces a biotin derivative on the surface of the beads. This proprietary mechanism allows users to achieve active release of the beads from target cells at any time within their process.

Based on a recommended 4-to-1 bead-to-target-cell ratio (not total nucleated cell number), one 15 mL vial can theoretically isolate up to 1.5 x 109 CD8+ T cells.

CTS Detachable Dynabeads also include a Drug Master File (DMF) to support cross-referencing in regulatory submissions prior to clinical trial and commercial manufacturing use.

Specifications

Concentration 4 x 108 beads/mL
Content And Storage CTS™ Detachable Dynabeads™ CD8, 15 mL.

Store at 2°C to 8°C with vial upright to keep beads in suspension.
Color Brown
Material Polystyrene
Product Type Cell Isolation Reagent
For Use With (Equipment) CTS™ DynaCellect™ Magnetic Separation System
Product Line CTS Detachable Dynabeads
Purity or Quality Grade GMP-grade
Quantity 15 mL vial
Shipping Condition Ships at 2°C to 8°C with wet (not frozen) gel packs. Vial must remain upright to keep beads in suspension.
Volume (Metric) 15 mL
Diameter (Metric) 4.5 μm
Show More Show Less

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.