Learn More
Gibco™ CTS™ Detachable Dynabeads™ 10X Release Buffer
Description
Increase flexibility in your cell therapy workflow with active bead release where it fits your process.
The Gibco CTS Detachable Dynabeads 10X Release Buffer is meant for use only with CTS Detachable Dynabeads magnetic beads. When used together, they empower users with a first-of-its-kind cGMP cell selection technology that has an active release mechanism for process development, clinical trial, and commercial manufacturing use.
Features of the CTS Detachable Dynabeads 10X Release Buffer (when used with CTS Detachable Dynabeads magnetic beads) include:
- Enables users with process flexibility by allowing active release of beads from target cells at any time
- Optimizes target cell purity, yield, activation profile, and desired phenotype thanks to the active release mechanism that gives users more control
- Provides an automation-friendly solution by working directly with the CTS DynaCellect Magnetic Separation System
- Is suitable for both autologous and allogeneic cell therapy manufacturing processes
CTS™ Detachable Dynabeads™ 10X Release Buffer is a concentrated biotin solution designed to enable active release of CTS™ Detachable Dynabeads™ directly in a culture vessel or intermediate bag supporting a closed, flexible and streamlined workflow.
This approach can help simplify workflows, reduce manual intervention and manufacturing costs, and enable bead release at a process stage that supports downstream operations such as genetic modification and bead removal.
Specifications
Specifications
| Concentration | 10X |
| For Use With (Application) | Cell Therapy |
| For Use With (Equipment) | CTS DynaCellect Magnetic Separation System |
| pH | 8.5 to 11.5 |
| Physical Form | Liquid |
| Product Line | Gibco |
| Quantity | 100 mL |
| Solution Type | Buffer |
| Packaging | Bottle |
| Sterility | Sterile |
| Show More |
For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.