Learn More
Gibco™ CTS™ AIM V™ SFM

Description
Gibco Cell Therapy Systems (CTS) AIM-V SFM is a defined, serum-free medium specifically designed for the proliferation and/or manipulation of T cells, dendritic cells, and other immune cells that is manufactured in compliance with current Good Manufacturing Practice (cGMP) standards.
CTS AIM V SFM contains L-glutamine, 50 μg/mL streptomycin sulfate, and 10 μg/mL gentamicin sulfate.
CTS quality
Gibco CTS products are manufactured at a site that uses methods and controls that conform to cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485-certified. Gibco CTS products follow USP 'ancillary materials for cell, gene, and tissue-engineered products' within the responsibilities applicable to a supplier.*
Accompanied by documentation to support your regulatory filing
Specific intended-use statements, full documentation traceability, and convenient access to our Drug Master File (DMF) are available.
*Other aspects of USP will be the responsibility of the end-user to assess. Thermo Fisher Scientific cannot fulfill USP in regards to application and therapy-specific aspects (e.g., residual assessment and removal of the Ancillary Material).
Specifications
Specifications
| Cell Type | Monocytes, Dendritic Cells, T Cells, Hybridomas, PBMCs, Fibroblasts, Macrophages, Myeloma Cells, Lymphocytes |
| Classification | Animal Origin |
| Form | Liquid |
| Product Type | Cell Culture Medium |
| Serum Level | Serum-free |
| Sterility | Sterile-filtered |
| For Use With (Application) | Cell and Gene Therapy Research, Development, and Manufacturing |
| Manufacturing Quality | ISO 13485, MDSAP, FDA-registered, 21 CFR 820 |
| Quantity | 1 L |
| Shipping Condition | Ambient |
| Show More |
Frequently Asked Questions (FAQs)
The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.
The difference is in the intended usage of the two media. AIM V Medium (Cat. Nos. 12055083, 12055091) is for research use only whereas CTS AIM V SFM (Cat. Nos. 0870112DK, 0870112BK) is for research use or manufacturing of cell, gene, or tissue-based products.
These two media have comparable performance and same cGMP quality. The main difference is that CTS AIM-V Medium (Cat. Nos. A3830801, A3830802, A4672701) does not contain phenol red and antibiotics. This medium formulation has been used as an ancillary reagent in a therapeutic cancer vaccine approved by FDA.
For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.