missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ AIM-V™ Medium, without phenol red, without antibiotics

Catalog No. A3830801
Encompass
Change view
Click to view available options
Quantity:
1 L
10 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A3830801 1 L
A3830802 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A3830801 Supplier Gibco™ Supplier No. A3830801

Cell Therapy Systems (CTS) AIM-V Medium is a defined, serum-free medium (SFM) for the proliferation and/or manipulation of T cells, dendritic cells, and other immune cells manufactured in compliance with cGMP.

Gibco Cell Therapy Systems (CTS) AIM-V Medium is a defined, serum-free medium (SFM) for the proliferation and/or manipulation of T cells, dendritic cells, and other immune cells manufactured in compliance with cGMP. It contains L-glutamine, but phenol red, gentamicin sulfate, and streptomycin sulfate have been removed to meet global regulatory requirements for ancillary materials used in cell therapy manufacturing.

CTS AIM-V Medium can be supplemented with human AB serum or CTS Immune Cell SR and various cytokines or growth factors depending on the application.

  • cGMP manufactured in compliance with 21 CFR part 820
  • Supports expansion of human peripheral blood T-cells including CAR T cells
  • Supports expansion of activated human dendritic cells and other immune cells
  • Proven clinical use as an ancillary reagent in Provenge™ (sipuleucel-T), the first therapeutic cancer vaccine approved by the FDA (1)
  • Adherence to supplier-related responsibilities of USP <1043>*

Closed systems compatible

For convenience, CTS AIM-V Medium is available in three formats: a 1000-mL bottle for small volume users, a 10-L bag for larger volume users, and a 2-L bioprocess container designed for users interested in a closed system workflow. The BPC chamber is constructed from Aegis 5-14 film and includes sterile, weldable DEPH-free PVC tubing with Luer lock and C-Flex™ tubing with MPC quick connector providing versatile connection options for your manufacturing process.

Security of supply

CTS AIM-V Medium is a defined serum-free formulation and can be manufactured at scale required to meet future clinical and commercial demand. Alternate manufacturing sites are available as further risk mitigation.

CTS quality

Gibco CTS products are manufactured at a site that uses methods and controls that conform to cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485-certified. Gibco CTS products follow USP <1043> 'ancillary materials for cell, gene, and tissue-engineered products' within the responsibilities applicable to a supplier.*

Accompanied by documentation to support your regulatory filing

Specific intended-use statements, full documentation traceability, and convenient access to our Drug Master File (DMF) are available.

Immune cell applications

CTS AIM-V Medium is cited in many peer-reviewed journals and used in multiple immune cell clinical applications such as activated dendritic cells. It supports expansion of T cells, T cell subsets, and virus-specific T cells. The medium can also be used to support natural killer cells combined with feeders. Each lot of CTS AIM-V SFM is pre-qualified using Jurkat cells, an immortalized human T lymphocyte line, to help ensure consistent performance.

*Other aspects of USP <1043> will be the responsibility of the end-user to assess. Thermo Fisher Scientific cannot fulfill USP <1043> in regards to application and therapy-specific aspects (e.g., residual assessment and removal of the AM).

1. Madan RA, Gulley JL. Sipuleucel-T: harbinger of a new age of therapeutics for prostate cancer. Expert Rev Vaccines. 2011 10(2):141-50.

Specifications

Cell Type Monocytes, Dendritic Cells, T Cells, Hybridomas, PBMCs, Fibroblasts, Macrophages, Myeloma Cells, Lymphocytes
Classification Animal Origin
Form Liquid
Product Type Cell Culture Medium
Serum Level Serum-free
Sterility Sterile-filtered
Manufacturing Quality ISO 13485, MDSAP, FDA-registered, 21 CFR 820
Quantity 1 L
Shipping Condition Ambient
Format Bottle
Content And Storage Store at 2–8°C and protect from light.
Show More Show Less
What is CTS?

The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems. Learn more here.

What is the difference between CTS AIM-V Medium and CTS AIM V SFM?

These two media have comparable performance and same cGMP quality. The main difference is that CTS AIM-V Medium (Cat. Nos. A3830801, A3830802, A4672701) does not contain phenol red and antibiotics. This medium formulation has been used as an ancillary reagent in a therapeutic cancer vaccine approved by FDA.

Is CTS AIM-V Medium, without phenol red, without antibiotics ready-to-use?

Yes, CTS AIM-V Medium, without phenol red, without antibiotics is ready-to-use. Additional supplementation with cytokines and growth factors may be required for certain types of cells. Human serum or CTS Immune Cell SR (Cat. Nos. A2596101 and A2596102) can be added to AIM-V medium to promote cell expansion.

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.