Learn More
Cellular Technology Ltd CTL TEST MEDIA 100ML DFS Item
Supplier:  Cellular Technology Ltd CTLT010
CTL-Test Medium, 100ml. CTL-Test Medium has been formulated to permit direct ex vivo testing of PBMC in T cell cytokine assays (ELISPOT, ELISA, CBA, CPA), while showing an equal or better performance than pre-screened, non-mitogenic serum batches. CTL-Test Medium contains injection grade water supplemented with amino acids, vitamins, inorganic salts, and human serum albumin (HSA) in a proprietary composition to provide a nutritionally complete and balanced environment for the PBMC. A variant of this medium, CTL-Test B, has been specially formulated for B cell ELISPOT assays. CTL-Test Medium is available in 100ml or 500ml bottles. The 100ml bottles are convenient for average-sized experiments, avoiding the need for sterile filtration, saving the cost of filters. Each batch of CTL-Test Medium is thoroughly tested for an equally high performance in ELISPOT assays as the previous batches, in addition to standard quality assurance procedures.
Catalog No. NC0482539
Only null left
Explore available promotions