missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ INS-1 832/13 Rat Insulinoma Cell Line

Catalog No. SCC207
Change view
Click to view available options
Quantity:
≥ 1 x 106 cells/vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCC207 ≥ 1 x 106 cells/vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCC207 Supplier MilliporeSigma™ Supplier No. SCC207

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

INS-1 832/13 rat insulinoma cell line is a useful model for insulin secretion regulation and pancreatic islet beta-cell function studies.

  • The rat insulinoma cell line INS-1 is a well-established model for studies of pancreatic islet beta-cell function (1); however, the GSIS response of INS-1 cells may decrease over time
  • The INS-1 832/13 cell line is a subclone of INS-1 that was selected for robust GSIS, producing and secreting both rat and human insulin
  • INS-1 832/13 harbors a human insulin expression cassette allowing for human insulin secretion to be maintained over extended passages with selection (2)
  • The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality
  • Cells are tested negative for infectious diseases by a Mouse/Rat Comprehensive CLEAR panel by Charles River Animal Diagnostic Services
  • Cells are verified to be of rat origin and negative for inter-species contamination from mouse, chinese hamster, Golden Syrian hamster, human and non-human primate (NHP) as assessed by a Contamination CLEAR panel by Charles River Animal Diagnostic Services
  • Cells are negative for mycoplasma contamination
  • Each vial contains ≥ 1X10⁶ viable cells

Specifications

Content And Storage Store in liquid nitrogen
Format Vial
For Use With (Application) INS-1 832/13 rat insulinoma cell line is a useful model for insulin secretion regulation and pancreatic islet beta-cell function studies.
Quantity ≥ 1 x 106 cells/vial
Source INS-1 832/13 is a derivative of INS-1 cells originally established from an x-ray induced insulinoma in rat (1). The INS-1 832/13 cell line is a subclone of INS-1 that was stably transfected with a CMV promoter-human insulin expression plasmid carrying a geneticin (G418)-resistance marker for selection (2)