missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CSF1 Split FISH Probe
Description
- The CSF1 Split FISH Probe is a dual-color, ready-to-use fluorescent probe designed for identifying gene splits or break-apart events.
- Chromosome Region: 1p13.3/1p13.3
- Fluorophores: Texas Red/FITC
- Application: Fluorescence in situ hybridization (FISH) analysis
Specifications
Specifications
| Content And Storage | Store at 4°C in the dark. |
| Description | DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe |
| Detection Method | Fluorescence |
| Format | Tube |
| Label or Dye | FITC |
| For Use With (Application) | FISH-Ce |
| Gene ID (Entrez) | 1435 |
| Includes | DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe |
| Product Type | FISH Probe |
| Quantity | 100 μL |
| Show More |