A temporary adhesive. These wash away adhesives are used as a temporary bond for holding delicate crystals, metallurgical specimens, glass components, and ceramic substrates for dicing, slicing, drilling and polishing. These materials adhere readily to metals, glass, ceramic and then can be washed after machining away using various solvents. Use as a water soluble temporary bond, for precise high purity work.
Color: White
Softening: 125F (52C)
Flow point: 130F(54C)
Viscosity: 500 cps