missing translation for 'onlineSavingsMsg'
Learn More

Electron Microscopy Sciences Crystalbond™ 555 Adhesive Stick, White 90 GR

Catalog No. 50997399
Encompass

A temporary adhesive. These wash away adhesives are used as a temporary bond for holding delicate crystals, metallurgical specimens, glass components, and ceramic substrates for dicing, slicing, drilling and polishing. These materials adhere readily to metals, glass, ceramic and then can be washed after machining away using various solvents. Use as a water soluble temporary bond, for precise high purity work.
Color: White
Softening: 125F (52C)
Flow point: 130F(54C)
Viscosity: 500 cps

Catalog No. 50997399 Supplier Electron Microscopy Sciences Supplier No. 5040101

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A temporary adhesive. These wash away adhesives are used as a temporary bond for holding delicate crystals, metallurgical specimens, glass components, and ceramic substrates for dicing, slicing, drilling and polishing. These materials adhere readily to metals, glass, ceramic and then can be washed after machining away using various solvents. Use as a water soluble temporary bond, for precise high purity work.
Color: White
Softening: 125F (52C)
Flow point: 130F(54C)
Viscosity: 500 cps