missing translation for 'onlineSavingsMsg'
Learn More

Crystal Violet Lactone 97.0+%, TCI America™

Catalog No. C074125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C074125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C074125G Supplier TCI America Supplier No. C074125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Heat and pressure sensitive dye for laboratory applications.

Chemical Identifiers

CAS 1552-42-7
Molecular Formula C26H29N3O2
Molecular Weight (g/mol) 415.54
MDL Number MFCD00070611
InChI Key IPAJDLMMTVZVPP-UHFFFAOYSA-N
Synonym 3,3-Bis(p-dimethylaminophenyl)-6-dimethylaminophthalide, 6-(Dimethylamino)-3,3-bis[p-(dimethylamino)phenyl]phthalide
PubChem CID 73773
IUPAC Name 6-(dimethylamino)-3,3-bis[4-(dimethylamino)phenyl]-1,3-dihydro-2-benzofuran-1-one
SMILES CN(C)C1=CC=C(C=C1)C1(OC(=O)C2=CC(=CC=C12)N(C)C)C1=CC=C(C=C1)N(C)C

Specifications

Melting Point 180°C
Quantity 25 g
Physical Form Crystal/Powder
Formula Weight 415.54
Percent Purity ≥97.0% (HPLC,T)
Grade Research
Chemical Name or Material Crystal Violet Lactone
EINECSNumber (5)-0129
TSCA Yes